Learn More
Corning™ Endothelial Cell Growth Supplement (ECGS)
GREENER_CHOICE
Ideal for endothelial cell culture
137.00 €
Specifications
Content And Storage 2° to 8°C
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11553520
GREENER_CHOICE
View Documents
Corning™
354006
15 mg
137.00 €
15mg
Description
Description
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
Specifications
Specifications
2° to 8°C
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : Stable for at least three months at 2° to 8°C (lyophilized) or = one month at -20°C (solubilized). Avoid multiple freeze/thaws.