Learn More
Lonza™ Endotoxin
High potency endotoxin intended for use in endotoxin removal system challenges, i.e., depyrogenation ovens, and other spiking studies.
Brand:  Lonza™ 193783
Product Code. 11600461
  • 872.00 € / Each
Explore more special offers
Description
Description
  • High potency
  • Does not contain filler
  • Can be used for oven validation studies
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1.25 million EU/vial
1.25 million EU/vial
Endotoxin removal
5 viales