Learn More
ENMD-1198, MedChemExpress
MedChemExpress ENMD-1198 (IRC-110160), an orally active microtubule destabilizing agent, is a 2-methoxyestradiol analogue with antiproliferative and antiangiogenic activity. ENMD-1198 is suitable for inhibiting HIF-1alpha and STAT3 in human HCC cells and leads to reduced tumor growth and vascularization.
Brand:  Medchem Express HY-16196
Encompass_Preferred
Product Code. 16481599
  • 697.95 € / 1mg
Explore more special offers
Description
Description
ENMD-1198 (IRC-110160), an orally active microtubule destabilizing agent, is a 2-methoxyestradiol analogue with antiproliferative and antiangiogenic activity. ENMD-1198 is suitable for inhibiting HIF-1alpha and STAT3 in human HCC cells and leads to reduced tumor growth and vascularization.
Specifications
Specifications
ENMD-1198
Brownish Yellow
C20H25NO2
IRC-110160
311.42
98.87%
Solid
864668-87-1
Research
1 mg
C[C@@]12C=CC[C@@]1([H])[C@]3([H])CCC4=C(C=C(OC)C(C(N)=O)=C4)[C@@]3([H])CC2
311.42
Research
Cancer-Kinase/protease