Learn More
Sartorius Entris™ II Advanced Precision Balances
GREENER_CHOICE
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
Brand:  Sartorius BCA1202I-1S
1769.74 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 16692444
  • 2185.00 € / Each
Explore more special offers
Description
Description
  • Entris™ II Advanced balances - Quality, Value, and Consistency
  • Accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in, real-time level support
  • Increases reliability of weighing results by using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE Certificate, CSA for Canada
With Internal Calibration
Intuitive Graphic Touch Technology
Glass, PBT, Stainless Steel
≤0.9 seconds
0.2 A
D-sub connector, 9-pin; USB UTL; USB Device
1200 g
10 mg
Electronic Precision Balance
100/240 V (± 10%)
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products