Learn More
Memmert™ Entry Port 14 mm for UF110plus
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF110plusD6
Product Code. 17598955
  • 221.00 € / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Entry port with 14 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
Specifications
UF110plus Oven
Entry Port
14 mm