Learn More
Memmert™ Entry Port 57 mm for UN450plus
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UN450plusF8
Product Code. 17580236
  • 303.00 € / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Entry port with 57 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
Specifications
UN450plus Oven
Entry Port
57 mm