Learn More
Memmert™ Entry Port Custom Position
Silicone Entry Port flap and stopper closure custom position TBC in the back
Brand:  Memmert™ IPP410ecoF7
 View more versions of this product
Product Code. 17140235
  • 467.00 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Individual position at the back (please, state location; not in combination with illumination units)
  • Moisture tight, can be closed by silicone stopper
Specifications
Specifications
Clear
IPP410eco
Silicone
40 mm