Learn More
Memmert™ Entry Port Left Centre/Centre
For introducing connections at the side. Moisture tight and can be closed by flap and silicone stopper
Brand:  Memmert™ IPP1060ecoF0
 View more versions of this product
Product Code. 17130065
  • 138.00 € / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Standard position left centre/centre
  • Not for model size 260 with light module
  • Not for model size 110 with light module
Specifications
Specifications
Moisture tight
IPP1060eco