Learn More
Memmert™ Entry Port Left Centre/Top
For introducing connections at the side and can be closed by flap
Brand:  Memmert™ HPP750ecoF1
 View more versions of this product
Product Code. 17179874
  • 240.00 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Standard position left centre/top
  • Not for modelsize 110 with light module
Specifications
Specifications
Clear
HPP750eco
23 mm