Learn More
Memmert™ Entry Port Rear (Individual Position)
Entry port with flap closure and custom position TBC in the back
Brand:  Memmert™ HPP110ecoF6
 View more versions of this product
Product Code. 17179694
  • 306.00 € / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Not in combination with illumination units
  • Moisture tight
  • Can be closed by flap and silicone stopper
Specifications
Specifications
Clear
HPP110eco
23 mm