Learn More
Memmert™ Entry Port Standard Position
Silicone Entry Port flap and stopper closure standard position in the back
Brand:  Memmert™ IPP1060ecoD8
 View more versions of this product
Product Code. 17100065
  • 362.00 € / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Standard position rear (not in combination with illumination units)
  • Moisture tight, can be closed by silicone stopper
Specifications
Specifications
Clear
IPP1060eco
Silicone
40 mm