missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Scienceware™ Enviro-Safe Thermometer
Shop All SP Scienceware Products
Click to view available options
Temperature Range (Metric):
-20°C to +150°C
Unit Size:
Each
Description
Enviro-Safe™ thermometers are environmentally safe and filled with a biodegradable liquid that can be cleaned up with soap and water. They can be used in place of hazardous spirit or mercury filled thermometers. The glass is lead free and non-toxic and the packaging is recyclable.
- Non-toxic
- Non-hazardous
- Environmentally friendly
- Lead free glass
- Accuracy traceable to NIST and DKD (statement of accuracy included)
Specifications
Specifications
| Certifications/Compliance | NIST, DKD |
| Immersion Type | Partial |
| Length (Metric) | 305 mm |
| Product Type | Thermometer |
| Max. Temperature (Metric) | 150°C |
| Min. Temperature (Metric) | -20°C |
| Temperature Range (Metric) | -20°C to +150°C |
| Calibrated | Yes |
| Accuracy | ±1°C |
| Immersion Length | 76 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction