Learn More
3M™ Enviro Swab ENVSWB100
A pre-moistened swab, ready to use for collecting environmental samples on all types of food contact surfaces in the food industry.
Brand:  3M™ 7100130395
Encompass
Product Code. 17288540
  • POA / Pack of 100
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Ready-to-use swab reaches into corners and hard-to-reach areas for increased accuracy.
  • Tamper-evident band indicates if the swab has been used.
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
  • Resealable tube provides tight closure for sample transportation.
  • Self-contained swab and vial for increased productivity and efficiency.
Specifications
Specifications
Swab
100/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Enviro Swab ENVSWB100