missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Eosin-5-Maleimide

eosin-5-maleimide

Brand:  Molecular Probes™ E118

559.36 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


699.20 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 11504736

  • 756.00 € / 25mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Bioconjugates made with the thiol-reactive eosin-5-maleimide can be used as phosphorescent probes or as photosensitizers. With its high quantum yield (∼0.57) for singlet oxygen generation, eosin and its conjugates can be used as effective photooxidizers of diaminobenzidine (DAB) in high-resolution electron microscopy studies and in correlated fluorescence and electron microscopy applications.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 mg
Store in freezer (-5 to -30°C) and protect from light.
Room Temperature
Eosin
Maleimide
Thiol
Maleimide
Classic Dyes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3080104 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberE118
Lot Number2978445 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog NumberE118
Lot Number2850246 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog NumberE118
Lot Number2811986 Certificate TypeCertificate of Analysis Date29/09/2023 Catalog NumberE118
Lot Number2519446 Certificate TypeCertificate of Analysis Date11/11/2022 Catalog NumberE118
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Eosin-5-Maleimide > 25mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.