Learn More
Eosin B, Certified, Pure, Thermo Scientific Chemicals
29.95 € - 84.72 €
Chemical Identifiers
CAS | 548-24-3 |
---|---|
Molecular Formula | C20H6Br2N2Na2O9 |
MDL Number | MFCD00005041 |
Synonym | 4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, Eosin Bluish |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10404891
|
Thermo Scientific Acros
409420250 |
25 g | Glass bottle |
29.95 €
25g |
In Stock
Log in to see stock. |
||||
10404315
|
Thermo Scientific Acros
409421000 |
100 g | Glass bottle |
84.72 €
100g |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
548-24-3 | |
C20H6Br2N2Na2O9 | |
MFCD00005041 | |
4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, Eosin Bluish |
Specifications
548-24-3 | |
85% min. | |
MFCD00005041 | |
15, 3657 | |
Authentic | |
624.06 | |
Brown to Purple/Red | |
Eosin B, Pure |
0.96 to 1.22 | |
C20H6Br2N2Na2O9 | |
19, 233 | |
4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, Eosin Bluish | |
511 to 520nm (in water + 1% Na2CO3) | |
Pure | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.