Learn More
Thermo Scientific™ EPD Electronic Personal Dosimeters
This product has been discontinued and replaced with the EPD TruDose Electronic Personal Dosimeter.
2936.20 €
Specifications
Product Type Personal Dosimeter
View More Specs
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
12855592
View accessory products View Documents
Thermo Scientific™
EPD/1/62921/420
EPD-N2 Gamma-Neutron Dosimeter This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This product has been discontinued and replaced with the EPD TruDose Electronic Personal Dosimeter.
The EPD product line consists of:
Thermo Scientific EPD Mk2 Electronic Personal Dosimeter
  • Excellent response to gamma, beta, and X-radiation.
  • Single stand-alone dosimeter or part of a comprehensive dosimetry management system, such as webREMS or ViewPoint Enterprise.
  • Advanced radiological performance, 15keV to 10MeV, in a small, lightweight design.
  • Most complete dosimeter per IAEA Active Personal Dosimeter Intercomparison study IAEA-TECDOC-1564.
  • Multi-detector technology.
  • Excellent power management and battery monitoring.
  • Loud configurable audible alarm.
  • Excellent immunity to electromagnetic interference.
  • Easy-to-read display with optional backlight.
  • Rugged battery cap and enhanced clip retention.
  • Reliable LCD and case.
Thermo Scientific EPD N2 Electronic Personal Dosimeter
  • Excellent performance in mixed gamma/neutron fields and for low-dose measurements.
  • Advanced radiological performance, 20keV to 10MeV (photon), thermal (0.025eV) to 15MeV (neutron).
  • Multi-detector technology.
  • Direct display of Hp(10) for neutrons and for photons.
  • Outstanding immunity to electromagnetic interference.
  • Compatible with current or upgradeable Thermo Scientific EPD readers, software and accessories.
Thermo Scientific EPD G Electronic Personal Dosimeter
  • Excellent response to gamma and X-radiation.
  • Advanced radiological performance, 15keV to 10MeV, in a small, lightweight design.
  • Multi-detector technology.
  • Excellent power management and battery monitoring.
  • Loud configurable audible alarm.
  • Excellent immunity to electromagnetic interference.
  • Easy-to-read display with optional backlight.
  • Rugged battery cap and enhanced clip retention.
  • Reliable LCD and case.
  • Compatible with Thermo Scientific EPD teleadapter for wireless operations.
Thermo Scientific EMDS3 Maintenance Database System
  • Establishment of reference calibration data of new stock.
  • Allocation of stock to user definable work areas or locations.
  • Plain English fault diagnosis and reporting.
  • Tracking of units sent for repair or maintenance.
  • Interface to calibration checking procedures for external irradiators.
  • Fully automatic integration with the Thermo Scientific™ Auto Irradiator.
  • Production of calibration check reports.
Thermo Scientific EPD External Alarm Module
  • Three-way external alarm unit for EPD Mk2 Electronic Personal Dosimeter.
  • LED light from the EPD activates the vibration and audio alarm through fiber hook.
  • Easily attached, optional, audible earpiece alarm.
  • External audible alarm jack capability.
Thermo Scientific EPD Readers for EPD Mk2 Electronic Personal Dosimeter
IrDA Reader: Office environments and mobile applications
ACT-5 Desktop Reader: For demanding environments, and where only one specific EPD can be ‘seen’ by the reader
ACT-4 Desktop Reader with Keypad
Specifications
Specifications
Personal Dosimeter