Learn More
BRAND™ EPDM Rubber Conical Gaskets
Place between filter funnel and filter flask for a secure fit.
Brand:  BRAND™ 462827
 View more versions of this product
Product Code. 10101971
  • 92.85 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rubber (EPDM) construction
  • Conical shape
  • Provides a secure fit between filter funnel and filter flask
  • Supplied in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Used for Filter Funnels and Filter Flasks
10
EPDM
42 mm
25 mm
Rubber Conical Gasket