missing translation for 'onlineSavingsMsg'
Learn More

Epichlorohydrin, 99%, AcroSeal™, Thermo Scientific Chemicals

32.66 € - 43.62 €

Chemical Identifiers

CAS 106-89-8
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00005132
InChI Key BRLQWZUYTZBJKN-UHFFFAOYSA-N
Synonym epichlorohydrin, 2-chloromethyl oxirane, epichlorhydrin, 1-chloro-2,3-epoxypropane, glycidyl chloride, oxirane, chloromethyl, epichlorhydrine, 1,2-epoxy-3-chloropropane, 2,3-epoxypropyl chloride, chloropropylene oxide
PubChem CID 7835
ChEBI CHEBI:37144
IUPAC Name 2-(chloromethyl)oxirane
SMILES C1C(O1)CCl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10787898
View Documents Promotion Details
Thermo Scientific Acros
430711000
100 mL AcroSeal™ Glass Bottle
32.66 €
100mL
Please to purchase this item. Need a web account? Register with us today!
13478578
View Documents Promotion Details
Thermo Scientific Acros
430718000
800 mL AcroSeal™ Glass bottle
43.62 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-89-8
92.52
BRLQWZUYTZBJKN-UHFFFAOYSA-N
7835
2-(chloromethyl)oxirane
C3H5ClO
MFCD00005132
epichlorohydrin, 2-chloromethyl oxirane, epichlorhydrin, 1-chloro-2,3-epoxypropane, glycidyl chloride, oxirane, chloromethyl, epichlorhydrine, 1,2-epoxy-3-chloropropane, 2,3-epoxypropyl chloride, chloropropylene oxide
CHEBI:37144
C1C(O1)CCl

Specifications

106-89-8
1.1800g/mL
28°C
98.5% min. (GC)
1.4365 to 1.4385
15,3666
epichlorohydrin, 2-chloromethyl oxirane, epichlorhydrin, 1-chloro-2,3-epoxypropane, glycidyl chloride, oxirane, chloromethyl, epichlorhydrine, 1,2-epoxy-3-chloropropane, 2,3-epoxypropyl chloride, chloropropylene oxide
BRLQWZUYTZBJKN-UHFFFAOYSA-N
2-(chloromethyl)oxirane
7835
1.12 mPa.s (20°C)
99%
Epichlorohydrin
-57.0°C
115.0°C to 117.0°C
Authentic
C3H5ClO
MFCD00005132
1.18
Solubility in water: 60g/l (10°C). Other solubilities: misclible with alcohol, ether, chloroform,, trichloroethylene, carbon tetrachloride, acetone, and benzene
C1C(O1)CCl
92.52
CHEBI:37144
92.52
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-439-8

Product Identifier
  • Epichlorohydrin

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only