missing translation for 'onlineSavingsMsg'
Learn More

Epichlorohydrin, 99%

Product Code. 10615922 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
25mL
This item is not returnable. View return policy

Product Code. 10615922

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-89-8 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-89-8
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
InChI Key BRLQWZUYTZBJKN-UHFFFAOYSA-N
Synonym epichlorohydrin, 2-chloromethyl oxirane, epichlorhydrin, 1-chloro-2,3-epoxypropane, glycidyl chloride, oxirane, chloromethyl, epichlorhydrine, 1,2-epoxy-3-chloropropane, 2,3-epoxypropyl chloride, chloropropylene oxide
PubChem CID 7835
ChEBI CHEBI:37144
IUPAC Name 2-(chloromethyl)oxirane
SMILES C1C(O1)CCl

Specifications

Melting Point -57.0°C
Density 1.1800g/mL
Boiling Point 115.0°C to 117.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4365 to 1.4385
Quantity 1 L
Beilstein 17,6
Fieser 01,355; 04,222; 05,290
Merck Index 15,3666
Specific Gravity 1.18
Solubility Information Solubility in water: 60g/l (10°C). Other solubilities: misclible with alcohol, ether, chloroform,, trichloroethylene, carbon tetrachloride, acetone, and benzene
Viscosity 1.12 mPa.s (20°C)
Formula Weight 92.52
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Epichlorohydrin
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Epichlorohydrin
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
  • Carcinogenicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.