missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epigallocatechin, Thermo Scientific Chemicals

Product Code. 15414199
Click to view available options
Quantity:
10 mg

 

A natural product from green tea that induces apoptosis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 970-74-1
Color Pink
Molecular Formula C15H14O7
MDL Number MFCD00075939
Quantity 10 mg
Synonym --epigallocatechin, epigallocatechin, epigallocatechol, l-epigallocatechin, epi-gallocatechin, antiscurvy factor c2, --epigallocatechol, l-epigallocatechol, --3,3',4',5,5',7-flavanhexol, 3,3',4',5,5',7-flavanhexol
InChI Key XMOCLSLCDHWDHP-IUODEOHRSA-N
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)O
IUPAC Name (2R,3R)-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
Molecular Weight (g/mol) 306.27
PubChem CID 72277
ChEBI CHEBI:42255
Formula Weight 306.27
Physical Form Powder
Chemical Name or Material (-)-Epigallocatechin
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.