missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Epoch 2 Microplate Spectrophotometer
GREENER_CHOICE

Product Code. 17227809 Shop All Agilent BioTek Products
Click to view available options
Model:
EPOCH2NS
EPOCH2NSC
EPOCH2TS
EPOCH2TSC
Includes:
Cuvette Port
No Touchscreen, No Cuvette Port
Touchscreen only
Touchscreen; Cuvette port
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17227809

Brand: Agilent BioTek EPOCH2NSSN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For Research Use Only. Not for use in diagnostic procedures.

The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. Measurements can be performed in 6- to 384-well microplates, cuvets, and in microvolume samples with the available Take3 microvolume plate. The broad wavelength range enables applications from nucleic acid and protein quantification in the low UV to microbial growth assays higher wavelengths

Epoch 2 is easy to use and the optional touch screen interface makes it simple to choose predefined protocols or to define custom programs. Endpoint, kinetic, spectral scanning and well area scanning modes, plus incubation and shaking enable wide-ranging applications.

TRUSTED_SUSTAINABILITY

Especificaciones

Detector Type Photodiode
Light Source Xenon Flash Lamp
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Accuracy ±2 nm
Certifications/Compliance CE and TUV marked. RoHS compliant.
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Height (English) 22 in.
Hertz 50/60 Hz
Includes No Touchscreen, No Cuvette Port
Model EPOCH2NS
Wavelength Range 200 to 999 nm, 1 nm increments
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado