missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPOR Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
166.92 € - 475.74 €
Specifications
Antigen | EPOR |
---|---|
Concentration | 450 μg/mL |
Applications | Western Blot, ELISA |
Form | Liquid |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18037035
|
Proteintech
30766-1-AP-150UL |
150 μL |
475.74 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18036675
|
Proteintech
30766-1-AP-20UL |
20 μL |
166.92 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The EPOR antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EPOR. This antibody recognizes human, mouse antigen. The EPOR antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
EPOR | |
Western Blot, ELISA | |
Rabbit | |
2057 | |
IgG | |
Store at -20°C. Stable for one year after shipment. |
450 μg/mL | |
Liquid | |
Human, Mouse | |
EPO R, EPOR, erythropoietin receptor | |
Primary |