missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Epoxy-coated Steel Multi Roll Tape Dispenser
Scienceware™ Write-On™ Labeling Tape or any tape with a 7.6cm (3 in.) ID core will fit on the drum of this multi-roll dispenser
Brand: Bel-Art™ SP Scienceware™ H13468-0000
This item is not returnable.
View return policy
Description
- Large writing surface, 10.2 x 15.2cm (4 x 6 in.) for preparing tape labels
- Built in serrated cutting edge
- Includes disks to prevent rolls from binding against each other
Specifications
Tape Dispenser | |
11 cm | |
1 Ea. |
Red | |
7.6cm I.D. Tape |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel-Art™ SP Scienceware™ Epoxy-coated Steel Multi Roll Tape Dispenser > Size: 26 x 15 x 11cm
Spot an opportunity for improvement?Share a Content Correction