missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Epoxy Diagnostic Slides

These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use

Brand:  Epredia™ X1XER332L

76.56 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.


 View more versions of this product

Product Code. 12675187

  • 91.85 € / Pack of 100
Estimated Shipment: 05-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Binds and keeps liquids applied to the well area
  • Provides wettable wells for staining and growing cell cultures
  • Prevents cross-contamination
  • Hydrophobic coatings are very durable and withstand the strongest stains
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 mm
Epoxy
Diagnostic Slide
Blue
75 mm
1 mm
4
8 mm Well
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Epoxy Diagnostic Slides >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.