Learn More
Epredia™ Epoxy Diagnostic Slides
These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use
Brand:  Epredia™ X1XER307W
 View more versions of this product
Product Code. 16389485
  • 79.30 € / Pack of 100
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Binds and keeps liquids applied to the well area
  • Provides wettable wells for staining and growing cell cultures
  • Prevents cross-contamination
  • Hydrophobic coatings are very durable and withstand the strongest stains
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mm
Epoxy
Diagnostic Slide
White
75 mm
1 mm
5
8 mm Well
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Epredia™ Epoxy Diagnostic Slides >