missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Rotor for Centrifuge 5418 R

Eppendorf Rotor for Centrifuge 5418 R, precision redefined. Tailor your centrifugation with varied capacities and speeds. Accessorize with adaptors and buckets for versatility in molecular biology and life science applications.

Brand:  Eppendorf™ 5418707005

615.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 10690934

  • 650.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FA-45-18-11, Aerosol-tight - Includes rotor lid
Aerosol-tight Rotor
16,873 x g
Centrifuge 5418 R
Aluminum
18 x 1.5 to 2.0 mL Tubes
14,000 rpm
Rotor Lid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers