Learn More
Eppendorf™ twin.tec™ PCR Plates 96-well, 250 μL - PCR Plates
GREENER_CHOICE
Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
Brand:  Eppendorf™ 0030129326
 View more versions of this product
Product Code. 10176287
Quantity Price
1 92.85 € / Pack of 10
5 85.82 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
† Certificate, test procedures and detailed information available upon request.
Certifications
Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human DNA, DNase, RNase-free
8 x 12
10 x Individually Blister Wrapped
PCR Plate
Polypropylene, Polycarbonate
96
RT-PCR applications
PCR Clean and Free of Bacterial DNA (Non-sterile)
Yes
Standard
Autoclavable
Colorless
Semi-skirted
250 μL
Non-sterile
2250 x g
10 Pk.
Colorless