missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips
GREENER_CHOICE

Use these adapters with Eppendorf pipet tips. Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips are color-coded and autoclavable.

Brand:  Eppendorf™ 0030089723

11.64 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 12624607

  • 12.25 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use these adapters with Eppendorf pipet tips. Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips are color-coded and autoclavable.

  • Available in 25 and 50mL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
1 Piece
Light Gray
50 mL Eppendorf™ Quality Tip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.