missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Combitips™ Advanced Tips for Multipette™ Multi-dispensers - PCR Clean
GREENER_CHOICE

Product Code. 12614617 Shop All Eppendorf Products
Click to view available options
Volume (Metric):
0.1 mL
0.2 mL
0.5 mL
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12614617

Brand: Eppendorf™ 0030089820

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Ensures comfortable handling while preventing damage to gloves. Eppendorf; Combitips advanced™ PCR Pipet Tips meet the increased requirements of a modern laboratory.

Combitips advanced replaces Combitips plus. Functioning according to the positive displacement principle, these always dispense the correct desired volume, independent of the liquid density and its flow characteristics. Working with radioactive, aggressive, toxic or infectious liquids also becomes safer due to hermetically sealed piston preventing aerosol contamination. The new individual color coding facilitates quick identification of the desired tip.

  • Ideal for use with all Eppendorf Repeaters™
  • Nine sizes offer a maximum range of volumes with individual dispensing from 0.1 to 50 mL
  • Funnel geometry prevents damage to gloves and ensures comfortable handling
  • Elongated design allows users to empty all standard laboratory tubes completely
  • Allow for single-hand operation; easy-reach dispenser box conveniently dispenses through a chute
  • Perform automatic Combitip recognition and volume calculation
  • 3D design on the upper rim allows for easy recognition
  • Purity grade: PCR Clean Quality - portioning in 4 resealable bags (25 Tips per bag) ensures dust-free storage
  • Additional purity grades available: standard Eppendorf Quality or Biopur™ (Cat. Nos. 13-683-700 through -726)

Ideal for a range of applications, including ELISA, RIA, PCR set-up and general sample preparation or other serial pipetting tasks.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 mL
Sterility Non-sterile
Color Crystal
Autoclavable Not Autoclavable
Clean Claims PCR Clean (Non-sterile)
Format Bulk, Bagged
Tip Style Extended Length, Positive Displacement
Packaging 100 Pk. (4 Reclosable Bags x 25 Pk.)
Product Type Pipet Tips
Color Code Orange
Disposable Single-use
For Use With (Equipment) Eppendorf Multipette
Product Line CombiTips Advanced
Material Polypropylene
Fit for Purpose PCR Ready
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.