Learn More
Eppendorf™ Combitips™ Advanced Tips for Multipette™ Multi-dispensers - Eppendorf Quality
GREENER_CHOICE
Eppendorf's consumable of choice for all Eppendorf Multipette™ multi-dispensers. Available in 9 volume sizes (0.1 to 50 mL), Combitips advanced in 'Eppendorf Quality' dispense in increments as low as 100 mL and are ideally suited for a variety of standard lab applications. Also available in purity grades PCR clean, Biopur™ or Forensic DNA Grade.
Brand:  Eppendorf™ 0030089480
138.70 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
 View more versions of this product
Product Code. 12624597
  • 150.60 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Use these tips for comfortable handling while preventing damage to gloves. Eppendorf Combitips advanced™ Standard Pipetter Tips are optimized to meet the increased requirements of a modern laboratory, and offered in three different purity grades to support any application.
  • Combitips advanced are the ideal system components for all Eppendorf Multipettes
  • The 9 volume sizes (0.1 mL – 50 mL) offer a maximum range of dispensing volumes with increments as low as 100 nanoliters depending on the Combitip used
  • Elongated Combitips in volumes 2.5 mL, 5.0 mL and 10 mL make it possible to reach the bottom of the most common laboratory vessels
  • An individual color coding facilitates the quick identification of the desired Combitips
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL
Colorless
Eppendorf Quality (Non-sterile)
Extended Length, Positive Displacement
Pipette Tips
Single-use
Yes
Polypropylene
Non-sterile
Not Autoclavable
Bulk, Bagged
100 Pk. (4 Bags x 25 Pk.)
Light Gray
Eppendorf Multipette
CombiTips Advanced
Certified Quality