Learn More
Eppendorf™ Heat Sealer Film and Foils
GREENER_CHOICE
Create 100% hermetic seal, eliminate evaporation, and prevent cross-contamination
148.00 € - 208.00 €
Specifications
Autoclavable Autoclavable
Certified RNase/DNase-free Yes
View More Specs
Products 3
Product Code Brand For Use With (Application) Product Type Price Quantity & Availability  
Product Code Brand For Use With (Application) Product Type Price Quantity & Availability  
11518663
GREENER_CHOICE
View Documents
Eppendorf™
0030127870
High-strength adhesive that forms tight seals to minimize sample evaporation Storage Film
164.66 €
Pack of 100
In Stock
to see stock.
Add to basket
11508663
GREENER_CHOICE
View Documents
Eppendorf™
0030127854
Heat sealing PCR foil for sealing plates, especially recommended for low reaction volumes, best protection against evaporation PCR Foil
148.00 €
Pack of 100
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
11833432
GREENER_CHOICE
View Documents
Eppendorf™
0030127838
Hermetic sealing of PCR plates, especially recommended for low reaction volumes, best protection against evaporation PCR Film
208.00 €
Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
  • Requires Eppendorf heat sealer (Cat. No. E5390 000 016)
  • Transparent heat sealing film allows sample monitoring in the plates
  • Aluminum coated, heat-sealing foils are available in peel-off or pierceable versions
  • Foil stripper tool removes heat-sealed film and foils completely
  • Certified free of DNA, DNase, RNase, and PCR inhibitors
  • Autoclavable
Specifications
Specifications
Autoclavable
Yes