missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ HeatSealer S100 Accessory, Plate Adapters
Shop All Eppendorf Products
Click to view available options
Product Type:
High Profile Adapter
Low Profile Adapter
PCR Plate Adapter
Unit Size:
Each
Description
- Hermetic sealing of multiwell plates
- Eliminates evaporation in PCR, reducing cross contamination
- Seals 96- and 384-well plates
- Integrated thermostat prevents overheating
- Improved mechanics for even easier sealing
- PCR Plate Adapter included
Warranty
One year
Specifications
Specifications
| Wattage | 320 W |
| For Use With (Equipment) | HeatSealer S100 |
| Sealing Temperature Range | 170°C |
| Product Type | High Profile Adapter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction