missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes with Fixed Volume
GREENER_CHOICE

Product Code. 15713469 Shop All Eppendorf Products
Click to view available options
Volume (Metric):
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
Accuracy:
±0.6%
±0.7%
±0.8%
±1%
±1.2%
Color:
Blue
Gray
Yellow
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15713469

missing translation for 'mfr': Eppendorf™ 3124000075

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Resistant to chemical, thermal, and mechanical stress

  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display Calibration display and adjustment opening for specific liquids and volumes Ultra light PerfectPiston™ piston system constructed of chemically resistant Fortron™ Quick connection clip allows removal of lower part in two quick steps (not in 10μL pipetters) Spring loaded tip cone promotes tight fit to the tip Color indicates pipetter's volume

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±0.6%
Autoclavable Autoclavable
Color Yellow
For Use With 200 μL Pipette Tip
Type Fixed Volume Pipette
Volume (Metric) 100 μL
For Use With (Equipment) 200μL Pipette tip
Includes Spring loaded tip cone, ejector, yellow operating button
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.