Learn More
Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes with Fixed Volume
GREENER_CHOICE
Resistant to chemical, thermal, and mechanical stress
Brand:  Eppendorf™ 3124000121
189.99 EUR valid until 2025-12-31
Use promo code "24314" to get your promotional price.
 View more versions of this product
Product Code. 15763469
  • 205.82 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display Calibration display and adjustment opening for specific liquids and volumes Ultra light PerfectPiston™ piston system constructed of chemically resistant Fortron™ Quick connection clip allows removal of lower part in two quick steps (not in 10μL pipetters) Spring loaded tip cone promotes tight fit to the tip Color indicates pipetter's volume
TRUSTED_SUSTAINABILITY
Specifications
Specifications
±0.6%
Blue
Fixed Volume Pipette
1000μL Pipette tip
Autoclavable
1000 μL Pipette Tip
1000 μL
Spring loaded tip cone, ejector, blue operating button
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes with Fixed Volume > Volume: 1,000μL; Blue