missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals EPPS, 0.2M buffer soln., pH 8.5

Product Code. 15404079
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym HEPPS
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

Concentration 0.2 M
pH 8.5
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material EPPS buffer soln.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'