missing translation for 'onlineSavingsMsg'
Learn More
epsilon-Caprolactone, 99%, Thermo Scientific Chemicals Product Code.: 11488950

epsilon-Caprolactone, 99%, Thermo Scientific Chemicals

Product Code. 11488950
100 g
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11488950

Brand: Thermo Scientific Alfa Aesar A10299.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ε-Caprolactone is widely used as a monomer in the manufacturing of highly specialized polymers. It is mainly utilized as a precursor to caprolactam. It finds an application in the synthesis of polyglecaprone, which is used as a suture material in surgery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ε-Caprolactone is widely used as a monomer in the manufacturing of highly specialized polymers. It is mainly utilized as a precursor to caprolactam. It finds an application in the synthesis of polyglecaprone, which is used as a suture material in surgery.

Solubility
Miscible with water and most organic solvents.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Combustible liquid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-44-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00003267
InChI Key PAPBSGBWRJIAAV-UHFFFAOYSA-N
Synonym 6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptane
PubChem CID 10401
ChEBI CHEBI:17915
IUPAC Name oxepan-2-one
SMILES C1CCC(=O)OCC1

Specifications

Melting Point -2°C
Density 1.078
Boiling Point 235°C to 236°C
Flash Point 109°C (228°F)
Assay Percent Range 99%
Refractive Index 1.463
Quantity 100 g
Beilstein 106919
Solubility Information Miscible with water and most organic solvents.
Formula Weight 114.14
Percent Purity 99%
Chemical Name or Material epsilon-Caprolactone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247851 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberA10299.0E,A10299.22,A10299.0C,A10299.36
Lot Number10210621 Certificate TypeCertificate of Analysis Date08/05/2023 Catalog NumberA10299.22
Lot Number10239224 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10299.0E,A10299.22,A10299.0C,A10299.36
Lot Number10239969 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10299.0E,A10299.22,A10299.0C,A10299.36
Lot Number10238856 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10299.0E,A10299.22,A10299.0C,A10299.36
compliance-icons
Product Identifier
  • epsilon-Caprolactone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-Oxepanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
epsilon-Caprolactone, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.