missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf epT.I.P.S.™ Motion as SafeRack tips
GREENER_CHOICE

Product Code. p-200056774 Shop All Eppendorf Products
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
1.000 μL
50 μL
300 μL
Sterility:
Non-sterile
PCR Clean
Filtered:
Filtered
Non-filtered
missing translation for 'unitSize'
Pack of 960
This item is not returnable. View return policy

Product Code. 30019023

missing translation for 'mfr': Eppendorf 0030014600

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The new epMotion systems can reuse tips as e.g. economical choice for multiple wash steps of the same well.

  • Specially designed for use on the epMotion™ 5070 and 5075 Liquid Handling Workstation. epT.I.P.S. Motion SafeRack pipette tips, with and without filters
  • Developed in torsion-resistant, contamination-free racks that are recommended for the re-use of tips feature of epMotion 5073 systems.

Reuse Tips Feature:

  • The new epMotion systems can reuse tips as e.g. economical choice for multiple wash steps of the same well
  • Pipette tips will be placed back into the position of the box they were removed from
  • Returned tips will be reused within an application for the same source vessel
  • Individual compartments in rack to avoid cross-contamination between used tips
  • Available as standard and PCR clean quality for 50, 300, and 1000 μL
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Not Autoclavable
Format Box
Tip Style Standard, Straight
Product Type Robotic Tips
Disposable Single-use
For Use With (Equipment) Eppendorf epMotion
Graduated No
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.