missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ epTIPS™ Pipette Tip Reloads

Use this refill system for worry-free work in the molecular biology lab. Eppendorf™ epT.I.P.S.™ Pipette Tip Reloads allow contamination-free handling of the refill trays. Plastic lids protect the trays from direct contact during transfer to the working box.

Specifications

Autoclavable Autoclavable
Format Rack
For Use With (Equipment) Eppendorf™ Pipettor
Product Line epT.I.P.S.
View More Specs

Description

Description

Use this refill system for worry-free work in the molecular biology lab. Eppendorf™ epT.I.P.S.™ Pipette Tip Reloads allow contamination-free handling of the refill trays. Plastic lids protect the trays from direct contact during transfer to the working box.

  • Contamination free transfer of refill trays to the refill box (epT.I.P.S.™ box) in plastic covers
  • Racks can be autoclaved
  • Also available in PCR* clean quality, free of human DNA, DNase, RNase and PCR* inhibitors
  • 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack
Specifications

Specifications

Autoclavable
Rack
Eppendorf™ Pipettor
epT.I.P.S.
Videos
SDS
Documents

Documents

Product Certifications