missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ epT.I.P.S.™ Standard Pipette Tips
GREENER_CHOICE

Product Code. 10744587 Shop All Eppendorf Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
0.5 to 20 μL
100 to 5000 μL
1000 to 10,000 μL
2 to 200 μL
20 to 300 μL
250 to 2500 μL
50 to 1000 μL
50 to 1250 μL
Length (Metric):
34 mm
40 mm
46 mm
53 mm
55 mm
71 mm
76 mm
103 mm
115 mm
120 mm
165 mm
243 mm
Color:
Blue
Clear
Dark Gray
Dark Green
Green
Light Gray
Medium Gray
Orange
Purple
Red
Turquoise
Yellow
Packaging:
1000 Tips (2 Bags x 500 Tips)
1000 Tips (4 Bags x 250 Tips)
200 Tips (2 Bags x 100 Tips)
500 Tips (5 Bags x 100 Tips)
Unit Size:
Pack of 1000
Pack of 200
Pack of 500
This item is not returnable. View return policy

Product Code. 10744587

Brand: Eppendorf™ 0030000730

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Use these universal pipette tips with a wide variety of pipette models. Eppendorf™ epT.I.P.S.™ Standard Pipettor Tips feature optimal wetting properties, high transparency and special, certified purity grades.

  • Minimal attachment and ejection forces with the highest level of tightness
  • The ergonomically optimized cone geometry provides the optimal pipette fit on the Eppendorf pipette
  • Fine graduations make visual checks of the pipetting volume even easier
  • Fine retracted or extended tips simplify work with small volumes
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 to 1250 μL
Color Dark Green
Autoclavable Autoclavable
Length (English) 4.1 in.
Length (Metric) 103 mm
Format Bulk
Tip Style Universal
Packaging 1000 Tips (4 Bags x 250 Tips)
Quantity 1000 Pk.
Product Type Pipette Tips
For Use With (Equipment) Eppendorf
Graduated Yes
Product Line epT.I.P.S.
Material Polypropylene
Graduations Fine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.