missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EquiPhi29™ DNA Polymerase

Thermo Scientific EquiPhi29 DNA Polymerase is a proprietary phi29 DNA polymerase mutant developed through in vitro protein evolution.

90.25 €

Specifications

Product Type Stand-alone enzyme
For Use With (Application) MDA-WGA, RCA
Polymerase EquiPhi29 DNA Polymerase
Optimal Reaction Temperature 42°C
Shipping Condition Dry Ice
View More Specs

Products 1
Product Code Brand Quantity ProductName Price Quantity & Availability  
Product Code Brand Quantity ProductName Price Quantity & Availability  
15973292
View Documents
Thermo Scientific™
A39390
250 U
90.25 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific EquiPhi29 DNA Polymerase is a proprietary phi29 DNA polymerase mutant developed through in vitro protein evolution. This enzyme is significantly improved over phi29 DNA polymerase in protein thermostability, reaction speed, product yield, and amplification bias, while retaining all the benefits of the wild-type enzyme.

EquiPhi29 DNA Polymerase possesses strong strand displacement activity and 3'→5' proofreading exonuclease activity that acts preferentially on single-stranded DNA or RNA.

Features of EquiPhi29 DNA Polymerase
• Highest processivity and strand displacement activity—more than 70 kb long DNA stretches can be synthesized
• Low amplification bias
• Extremely high yields of amplified DNA, even from small amounts of template
• Highly accurate DNA synthesis in a short time 

Applications
EquiPhi29 DNA Polymerase may be used in a wide variety of applications including:
• Unbiased whole genome amplification (WGA)
• Rolling circle amplification (RCA)
• Protein-primed DNA amplification
• Cell-free cloning of lethal DNA
In situ genotyping with padlock probes
• RNA-primed DNA amplification

Notes
1. The addition of pyrophosphatase to the reaction mixture with EquiPhi29 DNA Polymerase may further enhance DNA synthesis
2. Because of the enzyme's 3'→5' proofreading exonuclease, 3’-modified primers are highly recommended.
3. EquiPhi29 DNA Polymerase can amplify DNA in a wide range of temperatures (30–45°C), with an optimal reaction temperature of 42°C.

Specifications

Specifications

Stand-alone enzyme
EquiPhi29 DNA Polymerase
Dry Ice
2 hours
MDA-WGA, RCA
42°C
10 U/μL
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.