Learn More
Erbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
109.55 € - 454.26 €
Identifiants chimiques
CAS | 14553-08-3 |
---|---|
Molecular Formula | C15H21ErO6 |
Molecular Weight (g/mol) | 464.59 |
MDL Number | MFCD00013490 |
InChI Key | FGQSJRDKBCVFHH-UHFFFAOYSA-N |
Synonym | erbium 2,4-pentanedionate, erbium iii acetylacetonate hydrate, erbium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy erbio oxy pent-3-en-2-one |
PubChem CID | 102601666 |
SMILES | [Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
11356668
|
Thermo Scientific Alfa Aesar
013214.04 |
2 g |
109.55 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366668
|
Thermo Scientific Alfa Aesar
013214.09 |
10 g |
454.26 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Erbium(III) 2,4-pentanedionate is commonly used in various catalysts and catalytic reagents for organic synthesis. It is also used for glass coloring, as an amplifier in fiber optics, and in lasers for medical and dental purposes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
14553-08-3 | |
464.59 | |
FGQSJRDKBCVFHH-UHFFFAOYSA-N | |
102601666 |
C15H21ErO6 | |
MFCD00013490 | |
erbium 2,4-pentanedionate, erbium iii acetylacetonate hydrate, erbium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy erbio oxy pent-3-en-2-one | |
[Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Spécification
14553-08-3 | |
99.9% (REO) | |
MFCD00013490 | |
Insoluble in water. | |
[Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
464.59 | |
464.59 | |
Hygroscopic |
Powder | |
C15H21ErO6 | |
erbium 2,4-pentanedionate, erbium iii acetylacetonate hydrate, erbium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy erbio oxy pent-3-en-2-one | |
FGQSJRDKBCVFHH-UHFFFAOYSA-N | |
erbium(3+) tris(2,4-dioxopentan-3-ide) | |
102601666 | |
Odorless | |
Erbium(III) 2, 4-pentanedionate |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.