Learn More
Erbium(III) acetate tetrahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
69.35 € - 615.06 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11378458
|
Thermo Scientific Alfa Aesar
040481.06 |
5 g |
69.35 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388458
|
Thermo Scientific Alfa Aesar
040481.14 |
25 g |
189.95 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368458
|
Thermo Scientific Alfa Aesar
040481.22 |
100 g |
615.06 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Erbium(III) acetate tetrahydrate participates in the synthesis and determination of crystal structures and stereochemical properties of lanthanide(III) complexes with ethylenediamine-N,N,N?,N?-tetraacetate. Participates in simple one-pot single-step synthesis of rare earth doped spherical mesoporous metal oxide nanospheres with upconversion emission ability in supercritical methanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15280-57-6 | |
MFCD00013034 |
Specifications
15280-57-6 | |
Er(OOCCH3)3·4H2O | |
Soluble in water. | |
99.9% | |
Odorless | |
2.114 g/mL | |
Erbium(III) acetate tetrahydrate |
Crystalline Aggregates | |
MFCD00013034 | |
416.45 (344.39 Anhydrous) | |
Reacton™ | |
Hygroscopic | |
(REO) |
Safety and Handling
missing translation for 'einecsNumber' : 247-067-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only