missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11433860
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10138-41-7
Molecular Formula Cl3Er
Molecular Weight (g/mol) 273.609
MDL Number MFCD00010989
InChI Key HDGGAKOVUDZYES-UHFFFAOYSA-K
Synonym erbium chloride, erbium trichloride, erbium chloride ercl3, ercl3, erbium iii chloride, anhydrous, erbium iii chloride, ultra dry, erbium iii chloride hexahydrate, erbium iii chloride, anhydrous, ?ercl3, erbium iii chloride, anhydrous, powder trace metals basis, erbium iii chloride, anhydrous, beads,-10 mesh trace metals basis
PubChem CID 66277
IUPAC Name trichloroerbium
SMILES Cl[Er](Cl)Cl

Specifications

Boiling Point 1500°C
Melting Point 774°C
Physical Form Beads
Quantity 10 g
Solubility Information Soluble in water,strong mineral acids. Slightly soluble in MeOH and THF.
Formula Weight 273.62
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Erbium (III) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.