missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

191.96 € - 687.42 €

Chemical Identifiers

CAS 10138-41-7
Molecular Formula Cl3Er
Molecular Weight (g/mol) 273.609
MDL Number MFCD00010989
InChI Key HDGGAKOVUDZYES-UHFFFAOYSA-K
Synonym erbium chloride, erbium trichloride, erbium chloride ercl3, ercl3, erbium iii chloride, anhydrous, erbium iii chloride, ultra dry, erbium iii chloride hexahydrate, erbium iii chloride, anhydrous, ?ercl3, erbium iii chloride, anhydrous, powder trace metals basis, erbium iii chloride, anhydrous, beads,-10 mesh trace metals basis
PubChem CID 66277
IUPAC Name trichloroerbium
SMILES Cl[Er](Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11308617
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035739.06
5 g
191.96 €
5g
Please to purchase this item. Need a web account? Register with us today!
11318617
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035739.14
25 g
687.42 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

10138-41-7
273.609
HDGGAKOVUDZYES-UHFFFAOYSA-K
66277
Cl[Er](Cl)Cl
Cl3Er
MFCD00010989
erbium chloride, erbium trichloride, erbium chloride ercl3, ercl3, erbium iii chloride, anhydrous, erbium iii chloride, ultra dry, erbium iii chloride hexahydrate, erbium iii chloride, anhydrous, ?ercl3, erbium iii chloride, anhydrous, powder trace metals basis, erbium iii chloride, anhydrous, beads,-10 mesh trace metals basis
trichloroerbium

Specifications

1500°C
10138-41-7
Cl3Er
erbium chloride, erbium trichloride, erbium chloride ercl3, ercl3, erbium iii chloride, anhydrous, erbium iii chloride, ultra dry, erbium iii chloride hexahydrate, erbium iii chloride, anhydrous, ?ercl3, erbium iii chloride, anhydrous, powder trace metals basis, erbium iii chloride, anhydrous, beads,-10 mesh trace metals basis
HDGGAKOVUDZYES-UHFFFAOYSA-K
trichloroerbium
66277
99.9%
Ampule
(REO)
774°C
Powder
MFCD00010989
Soluble in water,strong mineral acids. Slightly soluble in MeOH and THF.
Cl[Er](Cl)Cl
273.609
273.62
Odorless
Hygroscopic
Erbium(III) chloride, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 233-385-0

missing translation for 'rtecsNumber' : KD8575000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only