missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11371059
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Erbium(III) fluoride is an important colorant in glass manufacturing and porcelain enamel glazes. It is used to produce erbium metal. It is used as dopant in making optical products. It is highly employed as erbium doped fluoride fibers in optical communications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-83-3
Molecular Formula ErF3
Molecular Weight (g/mol) 224.25
MDL Number MFCD00016074
InChI Key QGJSAGBHFTXOTM-UHFFFAOYSA-K
IUPAC Name erbium(3+) trifluoride
SMILES [F-].[F-].[F-].[Er+3]

Specifications

Melting Point 1350°C
Physical Form Granules, Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 224.28
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 6.4 g/mL
Assay (REO)
Chemical Name or Material Erbium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'