missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erbium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Beschreibung
Used as laboratory chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation
Spezifikation
Melting Point | 130°C (in -4H2O) |
Physical Form | Crystalline |
Quantity | 100 g |
UN Number | UN1477 |
Solubility Information | Typically insoluble in water. |
IUPAC Name | erbium(3+);trinitrate;pentahydrate |
Formula Weight | 353.27 (Anhydrous) |
Percent Purity | 99.9% |
Sensitivity | Hygroscopic |
Assay | (REO) |
Mehr anzeigen |
Produktvorschläge
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z30K020 | Certificate of Analysis | 31/12/2024 | 012920.22,012920.14 |
U07K038 | Certificate of Analysis | 22/08/2024 | 012920.22,012920.14 |
U07K034 | Certificate of Analysis | 22/08/2024 | 012920.22,012920.14 |
R08K006 | Certificate of Analysis | 16/05/2024 | 012920.22,012920.14 |
Z06J014 | Certificate of Analysis | 01/01/2024 | 012920.22,012920.14 |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Erbium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur