missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10177413
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-51-3
Molecular Formula ErH10N3O14
Molecular Weight (g/mol) 443.35
MDL Number MFCD00149691
InChI Key LWHHUEHWVBVASY-UHFFFAOYSA-N
Synonym erbium iii nitrate pentahydrate, erbium trinitrate pentahydrate, erbium nitrate pentahydrate, acmc-20al2v, er.3no3.5h2o, erbium nitrate-water 1/3/5, erbium iii nitrate pentahydrate, reacton, erbium nitrate, pentahydrate,
PubChem CID 53249207
SMILES O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Pink
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 33 to 39% (Er)
Linear Formula Er(NO3)3·5H2O
Solubility Information Solubility in water: soluble
IUPAC Name erbium(3+);trinitrate;pentahydrate
Formula Weight 443.35
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Erbium(III) nitrate pentahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'