Learn More
Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
91.10 € - 252.00 €
Chemical Identifiers
CAS | 10031-51-3 |
---|---|
Molecular Formula | ErH10N3O14 |
Molecular Weight (g/mol) | 443.35 |
MDL Number | MFCD00149691 |
InChI Key | LWHHUEHWVBVASY-UHFFFAOYSA-N |
Synonym | erbium iii nitrate pentahydrate, erbium trinitrate pentahydrate, erbium nitrate pentahydrate, acmc-20al2v, er.3no3.5h2o, erbium nitrate-water 1/3/5, erbium iii nitrate pentahydrate, reacton, erbium nitrate, pentahydrate, |
PubChem CID | 53249207 |
SMILES | O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10217330
|
Thermo Scientific Acros
315140250 |
25 g | Plastic bottle |
91.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10177413
|
Thermo Scientific Acros
315141000 |
100 g | Plastic bottle |
252.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
10031-51-3 | |
443.35 | |
LWHHUEHWVBVASY-UHFFFAOYSA-N | |
53249207 |
ErH10N3O14 | |
MFCD00149691 | |
erbium iii nitrate pentahydrate, erbium trinitrate pentahydrate, erbium nitrate pentahydrate, acmc-20al2v, er.3no3.5h2o, erbium nitrate-water 1/3/5, erbium iii nitrate pentahydrate, reacton, erbium nitrate, pentahydrate, | |
O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
10031-51-3 | |
Crystalline Powder and/or Crystals | |
ErH10N3O14 | |
MFCD00149691 | |
Solubility in water: soluble | |
O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
443.35 | |
443.35 | |
Trace Metal Basis |
Pink | |
33 to 39% (Er) | |
Er(NO3)3·5H2O | |
erbium iii nitrate pentahydrate, erbium trinitrate pentahydrate, erbium nitrate pentahydrate, acmc-20al2v, er.3no3.5h2o, erbium nitrate-water 1/3/5, erbium iii nitrate pentahydrate, reacton, erbium nitrate, pentahydrate, | |
LWHHUEHWVBVASY-UHFFFAOYSA-N | |
erbium(3+) pentahydrate trinitrate | |
53249207 | |
99.9% | |
Erbium(III) nitrate pentahydrate |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only