missing translation for 'onlineSavingsMsg'
Learn More
Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11391049

Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11391049
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11391049

Brand: Thermo Scientific Alfa Aesar 011306.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Erbium(III) nitrate pentahydrate is used as a chemical intermediates, producing a catalyst and related compounds, and also primary and secondary intermediated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-51-3
Molecular Formula ErH10N3O14
Molecular Weight (g/mol) 443.35
MDL Number MFCD00149691
InChI Key LWHHUEHWVBVASY-UHFFFAOYSA-N
Synonym erbium iii nitrate pentahydrate, erbium trinitrate pentahydrate, erbium nitrate pentahydrate, acmc-20al2v, er.3no3.5h2o, erbium nitrate-water 1/3/5, erbium iii nitrate pentahydrate, reacton, erbium nitrate, pentahydrate,
PubChem CID 53249207
SMILES O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 130°C (in -4H2O)
Physical Form Crystalline Aggregates
Quantity 25 g
UN Number UN1477
Solubility Information Soluble in water.
IUPAC Name erbium(3+) pentahydrate trinitrate
Formula Weight 443.35 (353.27 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Erbium(III) nitrate pentahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW27K012 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog Number011306.22,011306.14
Lot NumberN06J073 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number011306.22,011306.14
Lot NumberX11J013 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number011306.22,011306.14
Lot NumberW11J037 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number011306.22,011306.14
Lot NumberU24J002 Certificate TypeCertificate of Analysis Date13/09/2023 Catalog Number011306.22,011306.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Erbium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.