missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) oxide, nanopowder, 99.5% (REO), Thermo Scientific Chemicals

Product Code. 11409870
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

S. A. 13 to 17m2/g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12061-16-4
Molecular Formula Er2O3
Molecular Weight (g/mol) 382.52
MDL Number MFCD00010991
InChI Key ZXGIFJXRQHZCGJ-UHFFFAOYSA-N
Synonym Erbia
IUPAC Name dierbium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Er+3].[Er+3]

Specifications

Melting Point 2400°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.5% (REO)
Solubility Information Soluble in acids
Formula Weight 382.56
Appearance Powder or lump
Density 8.64
Chemical Name or Material Erbium(III) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'